Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpind1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024382.1 | NP_077358.1 | 3 - 4 | 949 | 131 | |
XM_006248709.2 | XP_006248771.1 | 2 - 3 | 956 | 131 | ||
GenBank mRNA | BC091145.1 | - | 3 - 4 | 929 | 7372906 | 131 |
FQ218607.1 | - | 2 - 3 | 926 | 131 | ||
X74549.1 | - | 3 - 4 | 949 | 131 | ||
X74550.1 | - | 2 - 3 | 896 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |