Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr3c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012576.2 | NP_036708.2 | 3 - 4 | 1529 | 84 | |
XM_008772065.1 | XP_008770287.1 | 3 - 4 | 1530 | 84 | ||
XM_008772066.1 | XP_008770288.1 | 4 - 5 | 1580 | 84 | ||
GenBank mRNA | AB115420.1 | - | 3 - 4 | 1516 | 84 | |
AF455050.1 | - | 3 - 4 | 1520 | 84 | ||
AY066016.2 | - | 3 - 4 | 1524 | 84 | ||
M14053.1 | - | 3 - 4 | 1587 | 84 | ||
Y12264.1 | - | 3 - 4 | 1541 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Nuclear Receptor Rattus norvegicus Inventoried Probe spans exons |