Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epb41l4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107397.2 | NP_001100867.1 | 6 - 7 | 532 | 76 | |
XM_006254582.2 | XP_006254644.1 | 11 - 12 | 3804 | 76 | ||
XM_006254583.2 | XP_006254645.1 | 11 - 12 | 2910 | 76 | ||
XM_006254584.2 | XP_006254646.1 | 12 - 13 | 2556 | 76 | ||
XM_006254588.2 | XP_006254650.1 | 11 - 12 | 1110 | 76 | ||
XM_008772040.1 | XP_008770262.1 | 11 - 12 | 887 | 76 | ||
GenBank mRNA | BC168858.1 | - | 6 - 7 | 532 | 7097125 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Rattus norvegicus Probe spans exons |