Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100515.1 | NP_001093985.1 | 3 - 4 | 445 | 120 | |
GenBank mRNA | BC166810.1 | - | 3 - 4 | 445 | 8376140 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |