Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rprd1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008772003.1 | XP_008770225.1 | 6 - 7 | 939 | 68 | |
XM_214609.8 | XP_214609.6 | 6 - 7 | 939 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Rattus norvegicus 1700 Probe spans exons |