Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034995.1 | NP_001030167.1 | 2 - 3 | 225 | 70 | |
XM_006254636.2 | XP_006254698.1 | 2 - 3 | 319 | 70 | ||
XM_006254637.2 | XP_006254699.1 | 2 - 3 | 633 | 70 | ||
XM_006254639.2 | XP_006254701.1 | 2 - 3 | 321 | 70 | ||
XM_006254640.2 | XP_006254702.1 | 2 - 3 | 321 | 70 | ||
XM_008772069.1 | XP_008770291.1 | 2 - 3 | 185 | 70 | ||
GenBank mRNA | BC103648.1 | - | 2 - 3 | 225 | 7315158 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |