Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD735029 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199493.1 | NP_955787.1 | 10 - 11 | 1196 | 98 | |
XM_006254634.2 | XP_006254696.1 | 10 - 11 | 1303 | 98 | ||
GenBank mRNA | BC064660.1 | - | 10 - 11 | 1196 | 5623815 | 98 |
FQ211991.1 | - | 1252 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |