Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srp19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106157.2 | NP_001099627.1 | 4 - 5 | 306 | 76 | |
GenBank mRNA | BC167043.1 | - | 4 - 5 | 306 | 7319792 | 76 |
FQ233350.1 | - | 4 - 5 | 356 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons |