Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107108.3 | NP_001100578.1 | 9 - 10 | 1724 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |