Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ets2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107107.1 | NP_001100577.2 | 3 - 4 | 412 | 70 | |
XM_006248177.2 | XP_006248239.1 | 3 - 4 | 630 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |