Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ets2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107107.1 | NP_001100577.2 | 7 - 8 | 1038 | 81 | |
XM_006248177.2 | XP_006248239.1 | 7 - 8 | 1256 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |