Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099758.1 | NP_001093228.1 | 1 - 2 | 85 | 75 | |
GenBank mRNA | BF555819.1 | - | 1 - 2 | 92 | 75 | |
CF978233.1 | - | 1 - 2 | 50 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |