Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108103.1 | NP_001101573.1 | 10 - 11 | 2776 | 94 | |
XM_006242194.2 | XP_006242256.1 | 10 - 11 | 3165 | 94 | ||
XM_006242196.2 | XP_006242258.1 | 10 - 11 | 2772 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |