Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zcrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034940.1 | NP_001030112.1 | 4 - 5 | 364 | 78 | |
GenBank mRNA | BC099747.1 | - | 4 - 5 | 364 | 5598773 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |