Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pphln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108992.1 | NP_001102462.1 | 2 - 3 | 358 | 85 | |
XM_006242230.1 | XP_006242292.1 | 2 - 3 | 355 | 85 | ||
XM_006242231.1 | XP_006242293.1 | 2 - 3 | 355 | 85 | ||
XM_006242236.2 | XP_006242298.1 | 2 - 3 | 122 | 85 | ||
GenBank mRNA | AY724510.1 | - | 1 - 2 | 91 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |