Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hs3st5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106392.1 | NP_001099862.1 | 2 - 3 | 856 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Protease Rattus norvegicus Probe spans exons |