Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nt5dc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106393.1 | NP_001099863.1 | 7 - 8 | 830 | 149 | |
XM_008772980.1 | XP_008771202.1 | 7 - 8 | 846 | 149 | ||
XM_008772981.1 | XP_008771203.1 | 7 - 8 | 843 | 149 | ||
GenBank mRNA | BC162021.1 | - | 7 - 8 | 794 | 9098589 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |