Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfic | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031567.1 | NP_113755.1 | 4 - 5 | 705 | 110 | |
XM_006240850.2 | XP_006240912.1 | 4 - 5 | 791 | 110 | ||
GenBank mRNA | AB012233.2 | - | 4 - 5 | 705 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |