Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfic | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031567.1 | NP_113755.1 | 8 - 9 | 1267 | 151 | |
GenBank mRNA | AB012233.2 | - | 8 - 9 | 1267 | 151 | |
FQ213370.1 | - | 7 - 8 | 1192 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |