Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sirt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031649.1 | NP_001026819.1 | 7 - 8 | 768 | 68 | |
XM_006240896.1 | XP_006240958.1 | 8 - 9 | 925 | 68 | ||
XM_006240897.2 | XP_006240959.1 | 7 - 8 | 1394 | 68 | ||
XM_008765091.1 | XP_008763313.1 | 7 - 8 | 1419 | 68 | ||
XM_008765092.1 | XP_008763314.1 | 8 - 9 | 1164 | 68 | ||
GenBank mRNA | BC098923.1 | - | 7 - 8 | 768 | 7461534 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |