Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cactin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081447.1 | NP_001074916.1 | 8 - 9 | 1809 | 106 | |
XM_008765124.1 | XP_008763346.1 | 8 - 9 | 2913 | 106 | ||
XM_008765125.1 | XP_008763347.1 | 8 - 9 | 2913 | 106 | ||
XM_008765126.1 | XP_008763348.1 | 8 - 9 | 2300 | 106 | ||
XM_008765127.1 | XP_008763349.1 | 8 - 9 | 1984 | 106 | ||
GenBank mRNA | AY934505.1 | - | 8 - 9 | 1809 | 106 | |
BC166406.1 | - | 8 - 9 | 2439 | 9028620 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons |