Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhj1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108072.2 | NP_001101542.1 | 5 - 6 | 434 | 85 | |
GenBank mRNA | BC168179.1 | - | 5 - 6 | 434 | 9092463 | 85 |
FQ230285.1 | - | 5 - 6 | 703 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |