Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr2e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109614.1 | NP_001103084.1 | 3 - 4 | 506 | 71 | |
XM_006241009.2 | XP_006241071.1 | 3 - 4 | 635 | 71 | ||
XM_006241010.1 | XP_006241072.1 | 3 - 4 | 526 | 71 | ||
XM_006241011.1 | XP_006241073.1 | 3 - 4 | 553 | 71 | ||
GenBank mRNA | BC158787.1 | - | 3 - 4 | 464 | 7304442 | 71 |
FQ219797.1 | - | 3 - 4 | 438 | 71 | ||
FQ220392.1 | - | 3 - 4 | 436 | 71 | ||
FQ229401.1 | - | 3 - 4 | 553 | 71 | ||
FQ230290.1 | - | 3 - 4 | 441 | 71 | ||
FQ234550.1 | - | 3 - 4 | 486 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |