Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uqcr11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126097.2 | NP_001119569.1 | 2 - 3 | 243 | 90 | |
GenBank mRNA | BC157815.1 | - | 2 - 3 | 219 | 8366041 | 90 |
CB783645.1 | - | 2 - 3 | 221 | 90 | ||
FQ217807.1 | - | 2 - 3 | 253 | 90 | ||
FQ217927.1 | - | 2 - 3 | 214 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |