Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sctr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031115.1 | NP_112377.1 | 8 - 9 | 1064 | 60 | |
GenBank mRNA | BC081781.1 | - | 8 - 9 | 1022 | 7189388 | 60 |
X59132.1 | - | 8 - 9 | 1064 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |