Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfkfb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271063.1 | NP_001257992.1 | 4 - 5 | 431 | 62 | |
NM_001271064.1 | NP_001257993.1 | 3 - 4 | 305 | 62 | ||
NM_012621.4 | NP_036753.4 | 4 - 5 | 554 | 62 | ||
XM_006256790.2 | XP_006256852.1 | 3 - 4 | 2464 | 62 | ||
XM_006256791.2 | XP_006256853.1 | 4 - 5 | 639 | 62 | ||
XM_008773123.1 | XP_008771345.1 | 6 - 7 | 605 | 62 | ||
XM_008773124.1 | XP_008771346.1 | 6 - 7 | 601 | 62 | ||
XM_008773125.1 | XP_008771347.1 | 5 - 6 | 479 | 62 | ||
GenBank mRNA | AY707861.1 | - | 4 - 5 | 382 | 62 | |
GQ422136.1 | - | 3 - 4 | 285 | 62 | ||
X15579.1 | - | 4 - 5 | 411 | 62 | ||
X15580.1 | - | 3 - 4 | 1409 | 62 | ||
Y00702.1 | - | 4 - 5 | 554 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |