Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003752031.3 | XP_003752079.2 | 2 - 3 | 308 | 90 | |
XM_006256807.2 | XP_006256869.1 | 2 - 3 | 5075 | 90 | ||
XM_006256808.2 | XP_006256870.1 | 2 - 3 | 1161 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus Probe spans exons |