Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133567.2 | NP_598251.1 | 7 - 8 | 824 | 64 | |
GenBank mRNA | AF123047.1 | - | 7 - 8 | 751 | 64 | |
AJ007422.1 | - | 7 - 8 | 751 | 64 | ||
AJ007616.1 | - | 7 - 8 | 751 | 64 | ||
BC099775.1 | - | 7 - 8 | 824 | 7933317 | 64 | |
U51013.1 | - | 7 - 8 | 857 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |