Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdgfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006248918.2 | XP_006248980.1 | 6 - 7 | 1119 | 131 | |
XM_008768940.1 | XP_008767162.1 | 7 - 8 | 2718 | 131 | ||
GenBank mRNA | BC061731.1 | - | 6 - 7 | 1124 | 5598667 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |