Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_221917.8 | XP_221917.5 | 3 - 4 | 401 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |