Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crhr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022714.1 | NP_073205.1 | 4 - 5 | 641 | 69 | |
XM_006236556.2 | XP_006236618.1 | 5 - 6 | 631 | 69 | ||
XM_006236557.2 | XP_006236619.1 | 5 - 6 | 628 | 69 | ||
XM_006236558.1 | XP_006236620.1 | 4 - 5 | 659 | 69 | ||
GenBank mRNA | EF078964.1 | - | 4 - 5 | 426 | 69 | |
U16253.1 | - | 4 - 5 | 641 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Detect Genomic DNA Rattus norvegicus Probe spans exons |