Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007646.1 | NP_001007647.1 | 8 - 9 | 1485 | 85 | |
GenBank mRNA | BC081961.1 | - | 8 - 9 | 1485 | 7124723 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |