Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013209.1 | NP_001013227.1 | 1 - 2 | 87 | 93 | |
XM_006235863.2 | XP_006235925.1 | 1 - 2 | 133 | 93 | ||
XM_006235866.2 | XP_006235928.1 | 1 - 2 | 130 | 93 | ||
GenBank mRNA | BC078908.1 | - | 1 - 2 | 87 | 7113862 | 93 |
FQ214601.1 | - | 1 - 2 | 88 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |