Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013209.1 | NP_001013227.1 | 5 - 6 | 460 | 74 | |
XM_006235863.2 | XP_006235925.1 | 5 - 6 | 506 | 74 | ||
XM_006235864.2 | XP_006235926.1 | 6 - 7 | 563 | 74 | ||
XM_006235866.2 | XP_006235928.1 | 5 - 6 | 503 | 74 | ||
GenBank mRNA | BC078908.1 | - | 5 - 6 | 460 | 7113862 | 74 |
Gene Symbol: LOC690183 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001073579.4 | XP_001073579.1 | 1 | 454 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |