Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nom1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008762677.1 | XP_008760899.1 | 2 - 3 | 1119 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Probe spans exons |