Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006257307.2 | XP_006257369.1 | 5 - 6 | 1212 | 78 | |
XM_006257308.2 | XP_006257370.1 | 4 - 5 | 2433 | 78 | ||
XM_008773465.1 | XP_008771687.1 | 4 - 5 | 2467 | 78 | ||
XM_008773466.1 | XP_008771688.1 | 5 - 6 | 1205 | 78 | ||
XM_343823.6 | XP_343824.4 | 4 - 5 | 1141 | 78 | ||
GenBank mRNA | M63991.1 | - | 3 - 4 | 1033 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Rattus norvegicus 1700 Probe spans exons |