Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749116.3 | XP_003749164.1 | 4 - 5 | 634 | 99 | |
XM_008760475.1 | XP_008758697.1 | 3 - 4 | 368 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |