Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pi4k2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053735.1 | NP_446187.1 | 3 - 4 | 820 | 70 | |
GenBank mRNA | AY029199.1 | - | 3 - 4 | 820 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |