Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053526.2 | NP_445978.1 | 7 - 8 | 1310 | 61 | |
GenBank mRNA | AB042599.1 | - | 7 - 8 | 1319 | 61 | |
BC088124.1 | - | 7 - 8 | 1310 | 7326031 | 61 | |
FQ218535.1 | - | 2 - 3 | 124 | 61 | ||
FQ219519.1 | - | 5 - 6 | 595 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Rattus norvegicus Probe spans exons |