Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vwa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749146.3 | XP_003749194.3 | 4 - 5 | 428 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |