Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dntt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012461.1 | NP_001012479.1 | 5 - 6 | 770 | 141 | |
GenBank mRNA | BC089904.1 | - | 5 - 6 | 770 | 7384335 | 141 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |