Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109515.1 | NP_001102985.1 | 1 - 2 | 405 | 110 | |
XM_006231567.2 | XP_006231629.1 | 1 - 2 | 426 | 110 | ||
GenBank mRNA | BC166775.1 | - | 1 - 2 | 397 | 110 | |
FQ225266.1 | - | 1 - 2 | 158 | 110 | ||
FQ235317.1 | - | 1 - 2 | 158 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |