Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flvcr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008769906.1 | XP_008768128.1 | 6 - 7 | 1450 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |