Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pvrl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109076.1 | NP_001102546.1 | 1 - 2 | 549 | 121 | |
XM_006250256.2 | XP_006250318.1 | 1 - 2 | 518 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |