Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpgat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109376.2 | NP_001102846.1 | 2 - 3 | 449 | 156 | |
XM_008769875.1 | XP_008768097.1 | 3 - 4 | 507 | 156 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |