Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006250449.2 | XP_006250511.1 | 2 - 3 | 269 | 134 | |
XM_008769889.1 | XP_008768111.1 | 2 - 3 | 269 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |