Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casq1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159594.1 | NP_001153066.1 | 2 - 3 | 524 | 73 | |
GenBank mRNA | CA334109.1 | - | 2 - 3 | 524 | 73 | |
CB699232.1 | - | 2 - 3 | 111 | 73 | ||
FQ216689.1 | - | 2 - 3 | 101 | 73 | ||
FQ224875.1 | - | 3 - 4 | 351 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |