Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024882.1 | NP_001020053.1 | 9 - 10 | 1191 | 65 | |
XM_006250296.2 | XP_006250358.1 | 9 - 10 | 1286 | 65 | ||
GenBank mRNA | BC098058.1 | - | 9 - 10 | 1191 | 7457890 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |