Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lin9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001067807.5 | XP_001067807.3 | 9 - 10 | 1216 | 64 | |
XM_006250417.1 | XP_006250479.1 | 10 - 11 | 1055 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |