Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nenf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002851.1 | NP_001002851.1 | 1 - 2 | 186 | 80 | |
GenBank mRNA | AY623793.1 | - | 1 - 2 | 186 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |